Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197165.1 | NP_001184094.1 | 1 - 2 | 75 | 89 | |
XM_005615770.2 | XP_005615827.1 | 2 - 3 | 193 | 89 | ||
XM_005615771.1 | XP_005615828.1 | 2 - 3 | 77 | 89 | ||
XM_014114374.1 | XP_013969849.1 | 2 - 3 | 201 | 89 | ||
GenBank mRNA | HM135455.1 | - | 1 - 2 | 75 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Canis familiaris Probe spans exons |