Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197165.1 | NP_001184094.1 | 3 - 4 | 483 | 81 | |
XM_005615770.2 | XP_005615827.1 | 4 - 5 | 601 | 81 | ||
XM_005615771.1 | XP_005615828.1 | 4 - 5 | 485 | 81 | ||
XM_014114374.1 | XP_013969849.1 | 4 - 5 | 609 | 81 | ||
XM_022423001.1 | XP_022278709.1 | 3 - 4 | 454 | 81 | ||
GenBank mRNA | HM135455.1 | - | 3 - 4 | 483 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Canis familiaris Probe spans exons |