Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197167.1 | NP_001184096.1 | 4 - 5 | 734 | 68 | |
XM_005615763.2 | XP_005615820.1 | 5 - 6 | 905 | 68 | ||
XM_005615764.2 | XP_005615821.1 | 5 - 6 | 900 | 68 | ||
GenBank mRNA | HM135453.1 | - | 4 - 5 | 734 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Canis familiaris Probe spans exons |