Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FPGT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204810.1 | NP_001191739.1 | 3 - 4 | 340 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Canis familiaris Probe spans exons |