Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205162.1 | NP_001192091.1 | 2 - 3 | 1171 | 77 | |
XM_005619519.3 | XP_005619576.1 | 5 - 6 | 1507 | 77 | ||
XM_005619522.2 | XP_005619579.1 | 3 - 4 | 1304 | 77 | ||
XM_005619523.3 | XP_005619580.1 | 3 - 4 | 1359 | 77 | ||
XM_005619524.3 | XP_005619581.1 | 3 - 4 | 1274 | 77 | ||
XM_014113199.2 | XP_013968674.1 | 7 - 8 | 1629 | 77 | ||
XM_022418031.1 | XP_022273739.1 | 5 - 6 | 1499 | 77 | ||
XM_022418032.1 | XP_022273740.1 | 4 - 5 | 1415 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Canis familiaris Probe spans exons |