Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205191.1 | NP_001192120.1 | 5 - 6 | 934 | 70 | |
XM_022407408.1 | XP_022263116.1 | 5 - 6 | 1153 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Canis familiaris Probe spans exons |