Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2RY11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204441.1 | NP_001191370.1 | 2 | 1348 | 87 | |
GenBank mRNA | CO595827.1 | - | 1 | 68 | 87 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Canis familiaris Both primers and probe map within a single exon |