Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPLP0 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252137.1 | NP_001239066.1 | 5 - 6 | 700 | 88 | |
XM_005636025.3 | XP_005636082.1 | 6 - 7 | 855 | 88 | ||
XM_022410154.1 | XP_022265862.1 | 5 - 6 | 880 | 88 | ||
GenBank mRNA | CX986930.1 | - | 1 | 370 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Canis familiaris Probe spans exons |