Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252317.1 | NP_001239246.1 | 6 | 518 | 80 | |
NM_001252318.1 | NP_001239247.1 | 7 | 539 | 80 | ||
NM_001252319.1 | NP_001239248.1 | 7 | 536 | 80 | ||
NM_001252320.1 | NP_001239249.1 | 5 | 496 | 80 | ||
NM_001252321.1 | NP_001239250.1 | 5 | 499 | 80 | ||
NM_001252322.1 | NP_001239251.1 | 7 | 521 | 80 | ||
GenBank mRNA | DN270012.1 | - | 6 | 484 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Canis familiaris Both primers and probe map within a single exon |