Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPS9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252368.1 | NP_001239297.1 | 182 | 70 | ||
GenBank mRNA | DT541279.1 | - | 182 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Canis familiaris |