Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFX8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270887.1 | NP_001257816.1 | 4 - 5 | 409 | 116 | |
XM_022423975.1 | XP_022279683.1 | 7 - 8 | 646 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |