Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313851.1 | NP_001300780.1 | 2 - 3 | 617 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Canis familiaris Inventoried Probe spans exons |