Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wnt10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507601.1 | XP_003507649.1 | 1 - 2 | 116 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Probe spans exons |