Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fev | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507620.2 | XP_003507668.2 | 4 - 5 | 467 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Probe spans exons |