Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ybx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007653625.1 | XP_007651815.1 | 3 - 4 | 251 | 67 | |
GenBank mRNA | JI884482.1 | - | 3 - 4 | 251 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons |