Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbtps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244089.1 | NP_001231018.1 | 1 - 2 | 170 | 78 | |
GenBank mRNA | AF019611.1 | - | 1 - 2 | 170 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Protease Probe spans exons |