Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbtps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244089.1 | NP_001231018.1 | 2 - 3 | 325 | 72 | |
XM_007640118.1 | XP_007638308.1 | 3 - 4 | 340 | 72 | ||
GenBank mRNA | AF019611.1 | - | 2 - 3 | 325 | 72 | |
JI894194.1 | - | 1 - 2 | 154 | 72 | ||
JP050960.1 | - | 1 - 2 | 154 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Protease Probe spans exons |