Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246717.1 | NP_001233646.1 | 4 - 5 | 507 | 64 | |
GenBank mRNA | AJ428867.1 | - | 4 - 5 | 507 | 64 | |
JP061690.1 | - | 3 - 4 | 280 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Probe spans exons |