Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244007.1 | NP_001230936.1 | 2 - 3 | 509 | 78 | |
GenBank mRNA | AB020238.1 | - | 2 - 3 | 509 | 78 | |
AB043543.3 | - | 2 - 3 | 509 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons |