Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpsa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244033.1 | NP_001230962.1 | 1 | 181 | 77 | |
GenBank mRNA | L16589.1 | - | 1 | 181 | 77 | |
Z22749.1 | - | 1 | 181 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Both primers and probe map within a single exon |