Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244043.1 | NP_001230972.1 | 4 - 5 | 721 | 110 | |
XM_007642121.1 | XP_007640311.1 | 5 - 6 | 110 | |||
GenBank mRNA | AY392426.1 | - | 4 - 5 | 721 | 110 | |
JI872398.1 | - | 4 - 5 | 110 | |||
JP053273.1 | - | 4 - 5 | 110 | |||
L22552.1 | - | 3 - 4 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |