Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Icam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246690.1 | NP_001233619.1 | 4 - 5 | 1076 | 90 | |
GenBank mRNA | AF308456.1 | - | 4 - 5 | 1076 | 90 | |
JI866855.1 | - | 4 - 5 | 997 | 90 | ||
JP055129.1 | - | 4 - 5 | 997 | 90 |