Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246736.1 | NP_001233665.1 | 4 - 5 | 436 | 87 | |
GenBank mRNA | JI878699.1 | - | 4 - 5 | 451 | 87 | |
JP046069.1 | - | 4 - 5 | 451 | 87 | ||
U05959.1 | - | 4 - 5 | 436 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |