Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244018.2 | NP_001230947.1 | 4 - 5 | 534 | 82 | |
GenBank mRNA | FN825778.1 | - | 4 - 5 | 538 | 82 | |
JI894393.1 | - | 4 - 5 | 446 | 82 | ||
JP044451.1 | - | 4 - 5 | 446 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Transporter Recommended Assay Inventoried Probe spans exons |