Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244018.2 | NP_001230947.1 | 9 - 10 | 933 | 87 | |
GenBank mRNA | FN825778.1 | - | 9 - 10 | 937 | 87 | |
JP044451.1 | - | 9 - 10 | 845 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Transporter 3' Most Probe spans exons |