Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Insig2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244078.1 | NP_001231007.1 | 5 - 6 | 830 | 103 | |
XM_007640186.1 | XP_007638376.1 | 5 - 6 | 921 | 103 | ||
XM_007640187.1 | XP_007638377.1 | 4 - 5 | 542 | 103 | ||
GenBank mRNA | AF527629.1 | - | 5 - 6 | 830 | 103 | |
JI868716.1 | - | 5 - 6 | 837 | 103 | ||
JP061244.1 | - | 5 - 6 | 837 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Probe spans exons |