Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gna11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244051.1 | NP_001230980.1 | 2 - 3 | 479 | 88 | |
GenBank mRNA | DQ202706.1 | - | 2 - 3 | 479 | 88 | |
JI862434.1 | - | 2 - 3 | 341 | 88 | ||
JP039111.1 | - | 2 - 3 | 342 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Probe spans exons |