Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efemp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244849.1 | NP_001231778.1 | 3 - 4 | 653 | 80 | |
GenBank mRNA | AF046870.1 | - | 3 - 4 | 653 | 80 | |
JI889270.1 | - | 3 - 4 | 418 | 80 | ||
JP061104.1 | - | 3 - 4 | 418 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |