Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efemp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244849.1 | NP_001231778.1 | 7 - 8 | 1141 | 60 | |
GenBank mRNA | AF046870.1 | - | 7 - 8 | 1141 | 60 | |
JI889270.1 | - | 7 - 8 | 906 | 60 | ||
JP061104.1 | - | 7 - 8 | 906 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |