Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246795.1 | NP_001233724.1 | 1 - 2 | 65 | 77 | |
XM_007651404.1 | XP_007649594.1 | 2 - 3 | 884 | 77 | ||
GenBank mRNA | EF104644.1 | - | 1 - 2 | 65 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Apoptosis Probe spans exons |