Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdca4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498746.2 | XP_003498794.1 | 1 | 618 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Both primers and probe map within a single exon |