Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkg7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003512841.1 | XP_003512889.1 | 3 - 4 | 434 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Apoptosis 3' Most Detect Genomic DNA Recommended Assay Probe spans exons |