Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007654958.1 | XP_007653148.1 | 2 - 3 | 127 | 72 | |
GenBank mRNA | JP049450.1 | - | 1 - 2 | 100 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Probe spans exons |