Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prickle4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007649613.1 | XP_007647803.1 | 3 - 4 | 544 | 64 | |
XM_007649614.1 | XP_007647804.1 | 3 - 4 | 540 | 64 | ||
XM_007649615.1 | XP_007647805.1 | 3 - 4 | 375 | 64 | ||
XM_007649616.1 | XP_007647806.1 | 2 - 3 | 335 | 64 | ||
XM_007649617.1 | XP_007647807.1 | 2 - 3 | 331 | 64 | ||
XM_007649618.1 | XP_007647808.1 | 3 - 4 | 494 | 64 | ||
XM_007649619.1 | XP_007647809.1 | 2 - 3 | 285 | 64 | ||
XM_007649620.1 | XP_007647810.1 | 3 - 4 | 544 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Probe spans exons |