Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcn15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007607951.1 | XP_007606141.1 | 1 - 2 | 50 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Probe spans exons |