Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Magoh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495730.2 | XP_003495778.1 | 3 - 4 | 382 | 90 | |
GenBank mRNA | JI873895.1 | - | 3 - 4 | 356 | 90 | |
JP040696.1 | - | 3 - 4 | 356 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Probe spans exons |