Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003501104.1 | XP_003501152.1 | 2 - 3 | 175 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons |