Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007647209.1 | XP_007645399.1 | 1 - 2 | 193 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Protease Probe spans exons |