Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007647209.1 | XP_007645399.1 | 4 - 5 | 779 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Protease 3' Most Probe spans exons |