Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm2d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498230.2 | XP_003498278.1 | 2 - 3 | 413 | 109 | |
GenBank mRNA | JI884409.1 | - | 2 - 3 | 243 | 109 | |
JP058848.1 | - | 2 - 3 | 243 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Probe spans exons |