Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrtm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003505036.2 | XP_003505084.1 | 1 | 125 | 86 | |
XM_007645539.1 | XP_007643729.1 | 1 | 129 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Both primers and probe map within a single exon |