Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klk5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007650818.1 | XP_007649008.1 | 4 - 5 | 731 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Protease 3' Most Recommended Assay Probe spans exons |