Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plet1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498539.2 | XP_003498587.2 | 1 - 2 | 293 | 63 | |
GenBank mRNA | JI870244.1 | - | 1 - 2 | 259 | 63 | |
JP063116.1 | - | 1 - 2 | 257 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |