Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh1b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500086.1 | XP_003500134.1 | 1 | 820 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Both primers and probe map within a single exon |