Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrnb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007624845.1 | XP_007623035.1 | 5 - 6 | 733 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Ion Channel Probe spans exons |