Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hpcal4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007645566.1 | XP_007643756.1 | 1 - 2 | 158 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Probe spans exons |