Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spink8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007644785.1 | XP_007642975.1 | 3 - 4 | 293 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |