Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc16a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007643055.1 | XP_007641245.1 | 3 - 4 | 1154 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Transporter 3' Most Probe spans exons |