Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Resp18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007651227.1 | XP_007649417.1 | 2 - 3 | 160 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Probe spans exons |