Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbx21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003499596.2 | XP_003499644.1 | 3 - 4 | 352 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Probe spans exons |